2-Bromothiophenol 97%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: o-Bromothiophenol, 2-Bromobenzene-1-thiol, 2-Bromobenzenethiol, 1-Bromo-2-mercaptobenzene
221980010 221980100 221980500
ACRO221980010EA 31.8 EUR
ACRO221980010 ACRO221980100 ACRO221980500
2-Bromothiophenol 97%
2-Bromothiophenol
Formula: C₆H₅BrS
MW: 189.08 g/mol
Boiling Pt: 114…116 °C (10 mmHg)
Density: 1.604 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004827
CAS Number: 6320-02-1
EINECS: 228-665-4
UN: 2810
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6340 to 1.6370 (20°C, 589 nm)
GC ≥96.0 %