3-Bromothiophenol 95%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: m-Bromothiophenol, 3-Bromobenzenethiol, 1-Bromo-3-mercaptobenzene, 3-Bromobenzene-1-thiol
303710010 303710050
ACRO303710010EA 38.9 EUR
ACRO303710010 ACRO303710050
3-Bromothiophenol 95%
3-Bromothiophenol
Formula: C₆H₅BrS
MW: 189.08 g/mol
Boiling Pt: 121 °C (40 mmHg)
Density: 1.6 g/cm³
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00009603
CAS Number: 6320-01-0
EINECS: 228-664-9
UN: 2810
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6320 to 1.6350 (20°C, 589 nm)
GC ≥94 %