4-Bromothiophenol 95%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 4-Bromobenzenethiol, p-Bromothiophenol, 4-Bromobenzene-1-thiol, 1-Bromo-4-mercaptobenzene
107460250 107461000 107460050
ACRO107460250EA 85 EUR
ACRO107460250 ACRO107461000 ACRO107460050
4-Bromothiophenol 95%
4-Bromothiophenol
Formula: C₆H₅BrS
MW: 189.08 g/mol
Boiling Pt: 239
Melting Pt: 71…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00004845
CAS Number: 106-53-6
EINECS: 203-407-3
UN: 2811
ADR: 6.1 III
Order Now
Specification Test Results
Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 71 to 76 °C
GC ≥94 %
Specific gravity (20/20 °C) 0.819 to 0.824
Acid value ≤8 mg KOH/g