4-Bromothiophenol ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
4-Bromobenzenethiol, p-Bromothiophenol, 4-Bromobenzene-1-thiol, 1-Bromo-4-mercaptobenzene
A10555.09
A10555.18
A10555.30
A10555.09EA
44.7
EUR
A10555.09
A10555.18
A10555.30
4-Bromothiophenol ≥98%
4-Bromothiophenol
Purity: 98%
CAS No.: 106-53-6
Molecular Formula: C6H5BrS
Formula Weight: 189.08
Odour: Unpleasant
Melting Point: 72-78°
Boiling Point: 239°
Beilstein Reference: 507031
UN No.: 2923
MDL No.: MFCD00004845
Formula:
C₆H₅BrS Boiling Pt: 239 °C Melting Pt: 72…78 °C Density: 1.526 g/cm³ (83 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004845 CAS Number: 106-53-6 EINECS: 203-407-3 UN: 2923 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...