Benzaldehyde
Supplier: EHRENSTORFER GMBH
EHERCA10532000EA
31.6
EUR
EHERCA10532000
Benzaldehyde
Benzaldehyde
Formula:
C₆H₅CHO MW: 106,12 g/mol Boiling Pt: 179 °C (1013 hPa) Melting Pt: –26 °C Density: 1,049 g/cm³ (20 °C) Flash Pt: 64 °C |
MDL Number:
MFCD00003299 CAS Number: 100-52-7 UN: 1990 ADR: 9,III Merck Index: 13,01057 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...