o-Anisaldehyde 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Salicylaldehyde methyl ether, 2-Methoxybenzaldehyde, 2-Anisaldehyde
149220050 149221000 149225000
ACRO149220050EA 34.1 EUR
ACRO149220050 ACRO149221000 ACRO149225000
o-Anisaldehyde 98%
o-Anisaldehyde
Formula: C₈H₈O₂
Boiling Pt: 238 °C (1013 hPa)
Melting Pt: 34…40 °C
Density: 1,127 g/cm³ (25 °C)
Flash Pt: 117 °C
Storage Temperature: Ambient
MDL Number: MFCD00003308
CAS Number: 135-02-4
EINECS: 205-171-7
Order Now
Specification Test Results
Appearance Light yellow to light brown Crystals or solid or liquids
Infrared spectrum Conforms
Melting point 34 to 40 °C
GC ≥97.5 %
Specific optical rotation +19.0° to +21.0° (20 °C, 589 nm) (c=2 in ethanol)