3-Hydroxy-p-anisaldehyde ≥98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Isovanillin, Isovanicaline, 3-Hydroxy-4-methoxybenzaldehyde, Isovanillic aldehyde
A12866.06 A12866.14 A12866.22
A12866.06EA 23.8 EUR
A12866.06 A12866.14 A12866.22
3-Hydroxy-p-anisaldehyde ≥98%
3-Hydroxy-p-anisaldehyde
Purity: 98% CAS No.: 621-59-0 Molecular Formula: C8H8O3 Formula Weight: 152.15 Flash Point: 110°(230°F) Melting Point: 112-116° Boiling Point: 179°/15mm Beilstein Reference: 1073021 MDL No.: MFCD00003369
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 179 °C (15 mmHg)
Melting Pt: 112…116 °C
Density: 1.20
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003369
CAS Number: 621-59-0
EINECS: 210-694-9