3-Hydroxy-p-anisaldehyde

Supplier: Thermo Scientific

Synonyms: Isovanillin, Isovanicaline, 3-Hydroxy-4-methoxybenzaldehyde, Isovanillic aldehyde

SB01291.1 SB01291.10 SB01291.50
MAYBSB01291.1EA 0 EUR
MAYBSB01291.1 MAYBSB01291.10 MAYBSB01291.50
3-Hydroxy-p-anisaldehyde
3-Hydroxy-p-anisaldehyde
Formula: C₈H₈O₃
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 68…70 °C
Density: 1.401 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003369
CAS Number: 621-59-0
EINECS: 210-694-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR