Syringaldehyde 98%
Supplier: Thermo Scientific
Warning
Synonyms: 3,5-Dimethoxy-4-hydroxybenzaldehyde, 4-Hydroxy-3,5-dimethoxybenzaldehyde
132880050 132880250 132881000
ACRO132880050EA 27.1 EUR
ACRO132880050 ACRO132880250 ACRO132881000
Syringaldehyde 98%
Syringaldehyde
Formula: C₉H₁₀O₄
MW: 182.18 g/mol
Boiling Pt: 192…193 °C (14 mmHg)
Melting Pt: 110…113 °C
Density: 1.013 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00006943
CAS Number: 134-96-3
EINECS: 205-167-5
Merck Index: 12,09191
Order Now
Specification Test Results
Appearance Light yellow to beige to green to brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 110 to 114 °C
GC ≥97.5 %