3,4,5-Trimethoxybenzaldehyde
Supplier: Thermo Scientific
SB00348.1 SB00348.10 SB00348.50
MAYBSB00348.1EA 0 EUR
MAYBSB00348.1 MAYBSB00348.10 MAYBSB00348.50
3,4,5-Trimethoxybenzaldehyde
3,4,5-Trimethoxybenzaldehyde
Formula: C₁₀H₁₂O₄
MW: 196.2 g/mol
Boiling Pt: 311
Melting Pt: 72…75 °C
Density: 1.367 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003364
CAS Number: 86-81-7
EINECS: 201-701-6