3-Hydroxybenzaldehyde 97
Supplier: Thermo Fisher Scientific
Warning
A13541.14 A13541.22 A13541.36
A13541.14EA 25.5 EUR
A13541.14 A13541.22 A13541.36
3-Hydroxybenzaldehyde 97
3-Hydroxybenzaldehyde
Purity: 97% CAS No.: 100-83-4 Molecular Formula: C7H6O2 Formula Weight: 122.12 Melting Point: 102-105° Boiling Point: 191°/50mm MDL No.: MFCD00003368
Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 191 °C (50 mmHg)
Melting Pt: 102…105 °C
Density: 1.1299 g/cm³ (114 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003368
CAS Number: 100-83-4
EINECS: 202-892-9