4-Hydroxybenzaldehyde, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
144088-250G 144088-50G
SIAL144088-250GEA 84.9 EUR
SIAL144088-250G SIAL144088-50G
4-Hydroxybenzaldehyde, Sigma-Aldrich®
4-Hydroxybenzaldehyde
Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 310
Melting Pt: 116
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 174
Storage Temperature: Ambient
MDL Number: MFCD00006939
CAS Number: 123-08-0
EINECS: 204-599-1
Merck Index: 13,04836