3-Phenoxybenzaldehyde

Supplier: Thermo Scientific

Synonyms: 3-Phenoxybenzenecarbaldehyde, m-Phenoxybenzenecarbaldehyde

SB01101.1 SB01101EA SB01101.50
MAYBSB01101.1EA 0 EUR
MAYBSB01101.1 MAYBSB01101EA MAYBSB01101.50
3-Phenoxybenzaldehyde
3-Phenoxybenzaldehyde
Formula: C₁₃H₁₀O₂
Boiling Pt: 392 °C (1013 hPa)
Melting Pt: 13 °C
Density: 1.147 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003353
CAS Number: 39515-51-0
EINECS: 254-487-1
UN: 2810
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR