3-Aminobenzonitrile 99%
Supplier: Thermo Fisher Scientific
Warning
A13995.09 A13995.18
A13995.09EA 38.7 EUR
A13995.09 A13995.18
3-Aminobenzonitrile 99%
3-Aminobenzonitrile
Purity: 99% CAS No.: 2237-30-1 Molecular Formula: C7H6N2 Formula Weight: 118.14 Flash Point: 110°(230°F) Odour: Almond-like Boiling Point: 290° UN No.: 3439 MDL No.: MFCD00007756
Formula: C₇H₆N₂
Boiling Pt: 290 °C
Melting Pt: 51…53 °C
Density: 1.14 g/cm³ (20 °C)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007756
CAS Number: 2237-30-1
EINECS: 218-800-5
UN: 3439
ADR: 6.1,III