3-Aminobenzonitrile 99%
Supplier: Thermo Fisher Scientific
|
Danger
|
173040250
173040050
173041000
ACRO173040250EA
82.4
EUR
ACRO173040250
ACRO173040050
ACRO173041000
3-Aminobenzonitrile 99%
3-Aminobenzonitrile
Formula:
C₇H₆N₂ Boiling Pt: 288…290 °C (1013 hPa) Melting Pt: 50…54 °C Density: 1,14 g/cm³ (20 °C) Flash Pt: 112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007756 CAS Number: 2237-30-1 EINECS: 218-800-5 UN: 3439 ADR: 6.1,III |
Specification Test Results
Appearance | Brown Crystalline solid |
Infrared spectrum | Conforms |
Melting point | 50 to 54 °C |
GC | ≥98.5 % |
Titration with HClO4 | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...