4-Aminobenzonitrile 98%
Supplier: Thermo Fisher Scientific
Danger
A13794.09 A13794.18 A13794.30
A13794.09EA 41 EUR
A13794.09 A13794.18 A13794.30
4-Aminobenzonitrile 98%
4-Aminobenzonitrile
Purity: 98% CAS No.: 873-74-5 Molecular Formula: C7H6N2 Formula Weight: 118.14 Melting Point: 83-87° Beilstein Reference: 774507 UN No.: 3439 MDL No.: MFCD00007821
Formula: C₇H₆N₂
Boiling Pt: 167 °C (1.1 mmHg)
Melting Pt: 83…87 °C
Density: 1.274 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007821
CAS Number: 873-74-5
EINECS: 212-850-1
UN: 3439
ADR: 6.1,III