3-Nitrobenzonitrile

Supplier: Thermo Scientific
Danger

Synonyms: m-Nitrobenzonitrile

SB00868.1 SB00868.10 SB00868.50
MAYBSB00868.1EA 0 EUR
MAYBSB00868.1 MAYBSB00868.10 MAYBSB00868.50
3-Nitrobenzonitrile
3-Nitrobenzonitrile
Formula: C₇H₄N₂O₂
MW: 148.12 g/mol
Boiling Pt: 258 °C (1013 hPa)
Melting Pt: 110 °C
Density: 0.33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007194
CAS Number: 619-24-9
EINECS: 210-587-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR