3,5-Dimethoxybenzonitrile ≥98%
Supplier: Thermo Scientific
Danger
B22989.06 B22989.14
B22989.06EA 55 EUR
B22989.06 B22989.14
3,5-Dimethoxybenzonitrile ≥98%
3,5-Dimethoxybenzonitrile
Purity: 98% CAS No.: 19179-31-8 Molecular Formula: C9H9NO2 Formula Weight: 163.18 Melting Point: 87-89° UN No.: 3439 MDL No.: MFCD00001804
Formula: C₉H₉NO₂
MW: 163.18 g/mol
Boiling Pt: 283 °C (1013 hPa)
Melting Pt: 87…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00001804
CAS Number: 19179-31-8
EINECS: 242-858-0
UN: 3439
ADR: 6.1,III