Butyl benzoate 98+%
Supplier: Thermo Fisher Scientific
Warning
296400010 296400050 296402500
ACRO296400010EA 89.3 EUR
ACRO296400010 ACRO296400050 ACRO296402500
Butyl benzoate 98+%
Butyl benzoate
Formula: C₁₁H₁₄O₂
Boiling Pt: 249…251 °C (1013 hPa)
Melting Pt: –22 °C
Density: 1 g/cm³
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00009439
CAS Number: 136-60-7
EINECS: 205-252-7
Merck Index: 13,01551
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4960 to 1.4990 (20°C, 589 nm)
GC ≥98.0 %