Benzoic acid, Sigma-Aldrich®
Supplier: Merck
Danger
MQ200
18102-1KG-R 18102-2.5KG-R 18102-500G-R 18102-6X1KG-R 18102-25KG-R
SIAL18102-1KG-REA 62.9 EUR
SIAL18102-1KG-R SIAL18102-2.5KG-R SIAL18102-500G-R SIAL18102-6X1KG-R SIAL18102-25KG-R
Benzoic acid, Sigma-Aldrich®
Benzoic acid
Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 249
Melting Pt: 121…123 °C
Density: 1.316 g/cm³ (20 °C)
Flash Pt: 121 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002398
CAS Number: 65-85-0
EINECS: 200-618-2
REACH: 01-2119455536-33
Merck Index: 13,01092
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d561f311b11ea8ab2c8f07253e6748e588eb71aa&vid=dcf5e45072cda5106216b497cd29716270ae772c&prid=web&clid=SAPDAM