Benzoic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
MQ200
33047-100G-R
33047-1KG-R
33047-250G-R
SIAL33047-100G-REA
38.3
EUR
SIAL33047-100G-R
SIAL33047-1KG-R
SIAL33047-250G-R
Benzoic acid, Sigma-Aldrich®
Benzoic acid
Formula:
C₇H₆O₂ MW: 122.12 g/mol Boiling Pt: 249 Melting Pt: 121…123 °C Density: 1.316 g/cm³ (20 °C) Flash Pt: 121 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00002398 CAS Number: 65-85-0 EINECS: 200-618-2 REACH: 01-2119455536-33 Merck Index: 13,01092 |