DSC calibration standard, benzoic acid
Supplier: LGC Standards
PROOLGC2606EA 478 EUR
PROOLGC2606
DSC calibration standard, benzoic acid
Standards Melting Point Standards
Enthalpy of fusion: 17.98 kJ/mol