4-Cyanobenzoic acid 99%
Supplier: Thermo Scientific
Warning
110730050 110730250
ACRO110730050EA 39.8 EUR
ACRO110730050 ACRO110730250
4-Cyanobenzoic acid 99%
4-Cyanobenzoic acid
Formula: C₈H₅NO₂
MW: 147.13 g/mol
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 217…222 °C
Density: 1,382 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002528
CAS Number: 619-65-8
EINECS: 210-606-9
Order Now
Specification Test Results
Appearance White to yellow to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 217 to 222 °C
HPLC ≥98.5 %
Titration by coupling ≥95 %