Anthranilic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Aminobenzoic acid
10680-100G 10680-25G
SIAL10680-100GEA 605 EUR
SIAL10680-100G SIAL10680-25G
Anthranilic acid, Sigma-Aldrich®
Anthranilic acid
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 312
Melting Pt: 144
Density: 1.362 g/cm³ (20 °C)
Flash Pt: 150
Storage Temperature: Ambient
MDL Number: MFCD00007712
CAS Number: 118-92-3
EINECS: 204-287-5
Merck Index: 13,00421