Anthranilic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
2-Aminobenzoic acid
M-Clarity™ quality level = MQ100
A89855-25G
A89855-100G
A89855-500G
SIALA89855-25GEA
29.7
EUR
SIALA89855-25G
SIALA89855-100G
SIALA89855-500G
Anthranilic acid, Sigma-Aldrich®
Anthranilic acid
Formula:
C₇H₇NO₂ MW: 137.14 g/mol Boiling Pt: 312 Melting Pt: 144 Density: 1.362 g/cm³ (20 °C) Flash Pt: 150 Storage Temperature: Ambient |
MDL Number:
MFCD00007712 CAS Number: 118-92-3 EINECS: 204-287-5 Merck Index: 13,00421 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...