4-Aminobenzoic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: Vitamin Bx, para-Aminobenzoic acid, PABA, Vitamin H1
M-Clarity™ quality level = MQ200
100536-50G 100536-1KG 100536-250G
SIAL100536-50GEA 30.8 EUR
SIAL100536-50G SIAL100536-1KG SIAL100536-250G
4-Aminobenzoic acid, Sigma-Aldrich®
4-Aminobenzoic acid
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 340 °C (1013 hPa)
Melting Pt: 188 °C
Density: 1.45 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00007894
CAS Number: 150-13-0
EINECS: 205-753-0
Merck Index: 13,00422