3,4-Diaminobenzoic acid
Supplier: Thermo Scientific
Warning
SB01435.1 SB01435.10 SB01435.50
MAYBSB01435.1EA 0 EUR
MAYBSB01435.1 MAYBSB01435.10 MAYBSB01435.50
3,4-Diaminobenzoic acid
3,4-Diaminobenzoic acid
Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Boiling Pt: 354
Melting Pt: 211
Density: 1.544 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007726
CAS Number: 619-05-6
EINECS: 210-577-2