4-Nitrobenzoic acid 99+%
Supplier: Thermo Fisher Scientific
Warning
128460010 128460250 128461000
ACRO128460010EA 86 EUR
ACRO128460010 ACRO128460250 ACRO128461000
4-Nitrobenzoic acid 99+%
4-Nitrobenzoic acid
Formula: C₇H₅NO₄
Boiling Pt: 359 °C (1013 hPa)
Melting Pt: 239…242 °C
Density: 1,65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007352
CAS Number: 62-23-7
EINECS: 200-526-2
Merck Index: 12,06686
Order Now
Specification Test Results
Appearance White to light yellow Powder or crystalline powder or crystals
Infrared spectrum Conforms
Melting point 238 to 242 °C
HPLC ≥99.0 %
Titration with NaOH 99.0 to 101.0 %