4-Nitrobenzoic acid, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ200

72910-1KG 72910-250G 72910-50G
SIAL72910-1KGEA 57.6 EUR
SIAL72910-1KG SIAL72910-250G SIAL72910-50G
4-Nitrobenzoic acid, Sigma-Aldrich®
4-Nitrobenzoic acid
Formula: C₇H₅NO₄
MW: 167.12 g/mol
Boiling Pt: 359
Melting Pt: 121
Density: 1.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007352
CAS Number: 62-23-7
EINECS: 200-526-2
Merck Index: 12,06686

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR