Storm water samplers, Nalgene®
Supplier: Thermo Fisher Scientific
1100-1000 1120-1000 1160-1000
NALG1100-1000EA 236 EUR
NALG1100-1000 NALG1120-1000 NALG1160-1000
Storm water samplers, Nalgene®
Samplers Fluid Samplers
Storm water samplers with HDPE bottle.
  • Leakproof when closure is applied
  • Device intended for one-time use
Suitable for collecting samples for inorganic analyses.
Certifikace: EPA NPDES MSGP.
Upozornení: Not for oil and grease sample collection.
*The product(s) on this page are outside the scope of the EU Directive 2019/904