Safety canisters
Supplier: BURKLE
2601-3005 2601-2010 2601-2020 2601-7010
BURK2601-3005EA 458.35 EUR
BURK2601-3005 BURK2601-2010 BURK2601-2020 BURK2601-7010
Safety canisters
Safety Cans
Space-saving safety canister, stainless steel, with screw cap, pressure-relief valve and PTFE seals for storage.
  • Protection from exploding through flame protection and excess pressure valve
  • High degree of chemical resistance
  • Pressure balance takes place from about 0,3 bar