Salicylic acid ≥99%
Supplier: Thermo Scientific
A12253.0EEA 99.4 EUR
A12253.0E A12253.36 A12253.30
Salicylic acid ≥99%
Salicylic acid
Purity: 99%
CAS No.: 69-72-7
Molecular Formula: C7H6O3
Formula Weight: 138.12
Flash Point: 157°(315°F)
Melting Point: 158-160°
Boiling Point: 211°/20mm
Beilstein Reference: 774890
Merck Reference: 14,8332
MDL No.: MFCD00002439
Formula: (HO)C₆H₄CO₂H
MW: 138,12 g/mol
Boiling Pt: 211 °C (20 mmHg)
Melting Pt: 158…160 °C
Density: 1,443
Flash Pt: 157 °C (315 °F)
MDL Number: MFCD00002439
CAS Number: 69-72-7
Merck Index: 14,8332
https://si.vwr.com/stibo/web/std.lang.all/99/00/749900.jpg