Salicylic acid, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2-Hydroxybenzoic acid, o-Hydroxybenzoic acid
84210-100G 84210-500G
SIAL84210-100GEA 27.2 EUR
SIAL84210-100G SIAL84210-500G
Salicylic acid, Sigma-Aldrich®
Salicylic acid
Formula: (HO)C₆H₄CO₂H
MW: 138.12 g/mol
Boiling Pt: 336 °C (1013 hPa)
Melting Pt: 157…159 °C
Density: 1.443 g/cm³ (20 °C)
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00002439
CAS Number: 69-72-7
EINECS: 200-712-3
Merck Index: 13,08411
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af1f695fa92656a135b5e1742f1cca28f4d255fb&vid=8f3aff6fb9a70e5dd194b8bc46019af74e48e90a&prid=web&clid=SAPDAM