Thiosalicylic acid 98%
Supplier: Thermo Scientific
Warning
Synonyms: 2-Thiobenzoic acid, 2-Mercaptobenzoic acid
138891000 138895000
ACRO138891000EA 79.4 EUR
ACRO138891000 ACRO138895000
Thiosalicylic acid 98%
Thiosalicylic acid
Formula: C₇H₆O₂S
MW: 154.19 g/mol
Boiling Pt: 230 °C (0.02 mmHg)
Melting Pt: 164…169 °C
Density: 1,489 g/cm³ (20 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00004836
CAS Number: 147-93-3
EINECS: 205-704-3
ADR: 9,
Merck Index: 12,09498
Order Now
Specification Test Results
Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 164 to 169 °C
Titration Argentometric 97.5 to 102.5 %
HPLC ≥97.5 %