4-Chloro-3-nitrobenzaldehyde
Supplier: Thermo Scientific
SB00934.1 SB00934.10 SB00934.50
MAYBSB00934.1EA 0 EUR
MAYBSB00934.1 MAYBSB00934.10 MAYBSB00934.50
4-Chloro-3-nitrobenzaldehyde
4-Chloro-3-nitrobenzaldehyde
Formula: C₇H₄ClNO₃
MW: 185.57 g/mol
Melting Pt: 57…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00007078
CAS Number: 16588-34-4
EINECS: 240-645-7