2-Chloroveratraldehyde
Supplier: Thermo Scientific
Synonyms:
2-Chloro-3,4-dimethoxybenzaldehyde
SB01329.1
SB01329.10
MAYBSB01329.1EA
0
EUR
MAYBSB01329.1
MAYBSB01329.10
2-Chloroveratraldehyde
2-Chloroveratraldehyde
Formula:
C₉H₉ClO₃ MW: 200.62 g/mol |
MDL Number:
MFCD00084942 CAS Number: 5417-17-4 EINECS: 226-515-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...