2-Chlorobenzonitrile ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: o-Chlorobenzonitrile

B20451.0B B20451.18 B20451.30
B20451.0BEA 310 EUR
B20451.0B B20451.18 B20451.30
2-Chlorobenzonitrile ≥98%
2-Chlorobenzonitrile

Purity: 98% CAS No.: 873-32-5 Molecular Formula: C7H4ClN Formula Weight: 137.57 Flash Point: 122°(252°F) Odour: Almond-like Boiling Point: 232° Beilstein Reference: 1363916 UN No.: 3439 MDL No.: MFCD00001779

Formula: C₇H₄ClN
MW: 137.57 g/mol
Boiling Pt: 232 °C
Melting Pt: 43…46 °C
Density: 1.19 g/cm³ (25 °C)
Flash Pt: 122 °C (252 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001779
CAS Number: 873-32-5
EINECS: 212-836-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR