3,5-Dichlorobenzonitrile
Supplier: Thermo Scientific
CD07878.1
CD07878.10
CD07878.50
MAYBCD07878.1EA
0
EUR
MAYBCD07878.1
MAYBCD07878.10
MAYBCD07878.50
3,5-Dichlorobenzonitrile
3,5-Dichlorobenzonitrile
Formula:
C₇H₃Cl₂N MW: 172.01 g/mol Boiling Pt: 269 °C (1013 hPa) Melting Pt: 66 °C |
MDL Number:
MFCD00001800 CAS Number: 6575-00-4 EINECS: 229-495-3 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...