4-Chlorobenzoic acid ≥98%
Supplier: Thermo Scientific
Warning
Synonyms: p-Chlorobenzoic acid, Chlorodracyclic acid, Chlorodracylic acid
A15135.0B A15135.30
A15135.0BEA 125 EUR
A15135.0B A15135.30
4-Chlorobenzoic acid ≥98%
4-Chlorobenzoic acid
Purity: 98+% CAS No.: 74-11-3 Molecular Formula: C7H5ClO2 Formula Weight: 156.57 Flash Point: 238°(460°F) Odour: Odourless Boiling Point: 275° Beilstein Reference: 907196 Merck Reference: 14,2125 MDL No.: MFCD00002531
Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 275 °C
Melting Pt: 239…243 °C
Density: 1.54
Flash Pt: 238 °C (460 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000258
CAS Number: 74-11-3
EINECS: 200-805-9
Merck Index: 14,2125