6-Chloroanthranilic acid

Supplier: Thermo Scientific
Warning

Synonyms: 2-Amino-6-chlorobenzoic acid

BTB04560.1 BTB04560.10 BTB04560.50
MAYBBTB04560.1EA 0 EUR
MAYBBTB04560.1 MAYBBTB04560.10 MAYBBTB04560.50
6-Chloroanthranilic acid
6-Chloroanthranilic acid
Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 153…154 °C
MDL Number: MFCD00051530
CAS Number: 2148-56-3
EINECS: 218-416-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR