4-Chloro-2-nitrobenzoic acid

Supplier: Thermo Scientific
Warning

SB00532.1 SB00532.10 SB00532.50
MAYBSB00532.1EA 0 EUR
MAYBSB00532.1 MAYBSB00532.10 MAYBSB00532.50
4-Chloro-2-nitrobenzoic acid
4-Chloro-2-nitrobenzoic acid
Formula: C₇H₄ClNO₄
Melting Pt: 139…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00007214
CAS Number: 6280-88-2
EINECS: 228-483-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR