3-Chloro-4-fluorobenzaldehyde

Supplier: Thermo Scientific
Warning

SEW00869.1 SEW00869.10
MAYBSEW00869.1EA 0 EUR
MAYBSEW00869.1 MAYBSEW00869.10
3-Chloro-4-fluorobenzaldehyde
3-Chloro-4-fluorobenzaldehyde
Formula: C₇H₄ClFO
MW: 158.56 g/mol
Boiling Pt: 66 °C (2 mmHg)
Melting Pt: 44…46 °C
MDL Number: MFCD00011735
CAS Number: 34328-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR