4-Chloro-2-fluorobenzaldehyde 99%
Supplier: Thermo Scientific
Warning
290620050 290620250
ACRO290620050EA 98 EUR
ACRO290620050 ACRO290620250
4-Chloro-2-fluorobenzaldehyde 99%
4-Chloro-2-fluorobenzaldehyde
Formula: C₇H₄ClFO
MW: 158.56 g/mol
Melting Pt: 58…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00143282
CAS Number: 61072-56-8
Order Now
Specification Test Results
Appearance White to yellow to light beige Powder or crystals
Infrared spectrum Conforms
Melting point 57 to 61 °C
GC ≥98.5 %