4-Chloro-2-fluorobenzaldehyde ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A16110.06
A16110.09
A16110.06EA
120
EUR
A16110.06
A16110.09
4-Chloro-2-fluorobenzaldehyde ≥98%
4-Chloro-2-fluorobenzaldehyde
Purity: 98%
CAS No.: 61072-56-8
Molecular Formula: C7H4ClFO
Formula Weight: 158.56
Melting Point: 58-60°
Beilstein Reference: 3537705
Formula:
C₇H₄ClFO Melting Pt: 58…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143282 CAS Number: 61072-56-8 |