4-Chloro-3-fluorobenzaldehyde 98%
Supplier: Thermo Fisher Scientific
Warning
294140050 294140250
ACRO294140050EA 50.9 EUR
ACRO294140050 ACRO294140250
4-Chloro-3-fluorobenzaldehyde 98%
4-Chloro-3-fluorobenzaldehyde
Formula: C₇H₄ClFO
MW: 158.56 g/mol
Melting Pt: 46…49 °C
Flash Pt: 96.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00143288
CAS Number: 5527-95-7
Order Now
Specification Test Results
Appearance White to yellow Crystalline solid
Infrared spectrum Conforms
GC ≥97.5 %