2-Chloro-4-fluorobenzonitrile

Supplier: Thermo Scientific

TL00226.1 TL00226.10 TL00226.50
MAYBTL00226.1EA 0 EUR
MAYBTL00226.1 MAYBTL00226.10 MAYBTL00226.50
2-Chloro-4-fluorobenzonitrile
2-Chloro-4-fluorobenzonitrile
Formula: C₇H₃ClFN
MW: 155.56 g/mol
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 63…65 °C
MDL Number: MFCD00042523
CAS Number: 60702-69-4
EINECS: 262-384-8
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR