2,4-Dichloro-5-fluorobenzoic acid
Supplier: Thermo Scientific
|
Warning
|
BTB15031.1
BTB15031.10
MAYBBTB15031.1EA
0
EUR
MAYBBTB15031.1
MAYBBTB15031.10
2,4-Dichloro-5-fluorobenzoic acid
2,4-Dichloro-5-fluorobenzoic acid
Formula:
C₇H₃Cl₂FO₂ Boiling Pt: 298 °C (1013 hPa) Melting Pt: 141…144 °C |
MDL Number:
MFCD00075281 CAS Number: 86522-89-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...