1,2-Dichloro-3-iodobenzene
Supplier: Thermo Scientific
Warning
TL00546.1 TL00546.10 TL00546.50
MAYBTL00546.1EA 0 EUR
MAYBTL00546.1 MAYBTL00546.10 MAYBTL00546.50
1,2-Dichloro-3-iodobenzene
1,2-Dichloro-3-iodobenzene
Formula: C₆H₃Cl₂I
MW: 272.9 g/mol
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: 35 °C
MDL Number: MFCD00001034
CAS Number: 2401-21-0
EINECS: 219-276-0