1-Chloro-3-iodobenzene

Supplier: Thermo Scientific

Synonyms: 3-Chloroiodobenzene

TL00516.1 TL00516.10 TL00516EE
MAYBTL00516.1EA 0 EUR
MAYBTL00516.1 MAYBTL00516.10 MAYBTL00516EE
1-Chloro-3-iodobenzene
1-Chloro-3-iodobenzene
Formula: C₆H₄ClI
MW: 238.45 g/mol
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: 54.5 °C
Density: 1,9255 g/cm³ (20 °C)
MDL Number: MFCD00001046
CAS Number: 625-99-0
EINECS: 210-920-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR