2,3,5,6-Tetrachloroaniline
Supplier: Thermo Scientific
MAYBBTB01374.1EA
0
EUR
MAYBBTB01374.1
CHMPBTB01374.250
2,3,5,6-Tetrachloroaniline
2,3,5,6-Tetrachloroaniline
Formula:
C₆H₃Cl₄N MW: 230,91 g/mol Boiling Pt: 298 °C (1013 hPa) Melting Pt: 110 °C |
MDL Number:
MFCD00007659 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...